missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Muscle Phosphofructokinase/PFKM/PFK-1 Antibody (2F11), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22615IR
This item is not returnable.
View return policy
Description
Muscle Phosphofructokinase/PFKM/PFK-1 Monoclonal antibody specifically detects Muscle Phosphofructokinase/PFKM/PFK-1 in Human samples. It is validated for Western Blot, ELISA
Specifications
| Muscle Phosphofructokinase/PFKM/PFK-1 | |
| Monoclonal | |
| DyLight 755 | |
| 6-phosphofructokinase, muscle type, EC 2.7.1,6-phosphofructo-1-kinase, EC 2.7.1.11, GSD7, MGC8699, PFK1, PFK-1, PFKA, PFK-A, PFKX, Phosphofructo-1-kinase isozyme A, Phosphofructokinase 1, phosphofructokinase, muscle, phosphofructokinase, polypeptide X, phosphofructokinase-M, phosphohexokinase | |
| Recombinant human Muscle Phosphofructokinase/PFKM/PFK-1(1-780aa) purified from E. coli | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 5213 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, ELISA | |
| 2F11 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction