missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Muscimol

Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
Description
Potent GABAA receptor agonist and partial GABAA-ρ receptor agonist. Inhibits memory retention and attenuates airway constriction in vivo.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 114.1 |
| Purity | >98% |
| Molecular Weight (g/mol) | 114.10 |
| Chemical Name or Material | Muscimol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction