missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Muscimol, Tocris Bioscience™
Potent GABAA agonist; also GABAA-ρ partial agonist
£80.99 - £561.54
Chemical Identifiers
CAS | 2763-96-4 |
---|---|
Molecular Formula | C4H6N2O2 |
Molecular Weight (g/mol) | 114.10 |
MDL Number | MFCD00057894 |
InChI Key | ZJQHPWUVQPJPQT-UHFFFAOYSA-N |
Synonym | muscimol, pantherine, agarin, agarine, 3 2h-isoxazolone, 5-aminomethyl, 5-aminomethyl-3-isoxyzole, pantherin, 5-aminomethyl-3-isoxazolol, 3-hydroxy-5-aminomethylisoxazole, 5-aminomethyl-3-hydroxyisoxazole |
PubChem CID | 4266 |
ChEBI | CHEBI:7035 |
IUPAC Name | 5-(aminomethyl)-2,3-dihydro-1,2-oxazol-3-one |
SMILES | NCC1=CC(=O)NO1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30001383
|
Tocris Bioscience
0289/1 |
1 mg |
£80.99
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30001074
|
Tocris Bioscience
0289/10 |
10 mg |
£151.18
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18762704
|
Tocris Bioscience
0289/50 |
50 mg |
£561.54
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent GABAA receptor agonist and partial GABAA-ρ receptor agonist. Inhibits memory retention and attenuates airway constriction in vivo.Chemical Identifiers
2763-96-4 | |
114.10 | |
ZJQHPWUVQPJPQT-UHFFFAOYSA-N | |
4266 | |
5-(aminomethyl)-2,3-dihydro-1,2-oxazol-3-one |
C4H6N2O2 | |
MFCD00057894 | |
muscimol, pantherine, agarin, agarine, 3 2h-isoxazolone, 5-aminomethyl, 5-aminomethyl-3-isoxyzole, pantherin, 5-aminomethyl-3-isoxazolol, 3-hydroxy-5-aminomethylisoxazole, 5-aminomethyl-3-hydroxyisoxazole | |
CHEBI:7035 | |
NCC1=CC(=O)NO1 |
Specifications
2763-96-4 | |
MFCD00057894 | |
ZJQHPWUVQPJPQT-UHFFFAOYSA-N | |
5-(aminomethyl)-2,3-dihydro-1,2-oxazol-3-one | |
4266 | |
114.1 | |
Muscimol |
C4H6N2O2 | |
muscimol, pantherine, agarin, agarine, 3 2h-isoxazolone, 5-aminomethyl, 5-aminomethyl-3-isoxyzole, pantherin, 5-aminomethyl-3-isoxazolol, 3-hydroxy-5-aminomethylisoxazole, 5-aminomethyl-3-hydroxyisoxazole | |
NCC1=CC(=O)NO1 | |
114.10 | |
CHEBI:7035 | |
>98% |
Safety and Handling
Recommended Storage : Store at Room Temperature