missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Muscarinic Acetylcholine Receptor M5/CHRM5 Antibody (5G3H8), Novus Biologicals™
Mouse Monoclonal Antibody
£217.00 - £389.00
Specifications
| Antigen | Muscarinic Acetylcholine Receptor M5/CHRM5 |
|---|---|
| Clone | 5G3H8 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000 |
| Applications | ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30029674
|
Novus Biologicals™
NBP3-27132-0.025ml |
0.025 mL |
£217.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30026006
|
Novus Biologicals™
NBP3-27132-0.1ml |
0.1 mL |
£389.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Muscarinic Acetylcholine Receptor M5/CHRM5 Monoclonal antibody specifically detects Muscarinic Acetylcholine Receptor M5/CHRM5 in Human samples. It is validated for ELISA,Western BlotSpecifications
| Muscarinic Acetylcholine Receptor M5/CHRM5 | |
| 1 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Mouse | |
| GPCR, Neuroscience | |
| PBS | |
| 1133 | |
| IgG1 | |
| Protein G purified |
| 5G3H8 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| cholinergic receptor, muscarinic 5, HM5, MGC41838, muscarinic acetylcholine receptor M5 | |
| Purified recombinant fragment of human Muscarinic Acetylcholine Receptor M5/CHRM5 expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel