missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Muscarinic Acetylcholine Receptor M5/CHRM5 Antibody (590123) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB10323JF585
This item is not returnable.
View return policy
Description
Muscarinic Acetylcholine Receptor M5/CHRM5 Monoclonal antibody specifically detects Muscarinic Acetylcholine Receptor M5/CHRM5 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| Muscarinic Acetylcholine Receptor M5/CHRM5 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| cholinergic receptor, muscarinic 5, HM5, MGC41838, muscarinic acetylcholine receptor M5 | |
| NS0 mouse myeloma cell line transfected with human CHRM5, Met1-Pro532, Accession # P08912 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 590123 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| GPCR, Neuroscience | |
| 1133 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction