missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Muscarinic Acetylcholine Receptor M3/CHRM3 Antibody (580011) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6378D
This item is not returnable.
View return policy
Description
Muscarinic Acetylcholine Receptor M3/CHRM3 Monoclonal antibody specifically detects Muscarinic Acetylcholine Receptor M3/CHRM3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF
Specifications
| Muscarinic Acetylcholine Receptor M3/CHRM3 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 580011 | |
| Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| cholinergic receptor, muscarinic 3, HM3, m3 muscarinic receptor, muscarinic acetylcholine receptor M3 | |
| NS0 mouse myeloma cell line transfected with human CHRM3, Accession # P20309 | |
| 0.1 mL | |
| GPCR | |
| 1131 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction