missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Muscarinic Acetylcholine Receptor M2/CHRM2 Rabbit anti-Human, Mouse, Rat, Clone: 3T5W8, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Muscarinic Acetylcholine Receptor M2/CHRM2 Monoclonal antibody specifically detects Muscarinic Acetylcholine Receptor M2/CHRM2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Muscarinic Acetylcholine Receptor M2/CHRM2 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 3T5W8 |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Formulation | PBS, 0.05% BSA, 50% glycerol, pH7.3 |
| Gene Alias | 7TM receptor, cholinergic receptor, muscarinic 2, FLJ43243, HM2, MGC120006, MGC120007, muscarinic acetylcholine receptor M2, muscarinic M2 receptor |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 100-200 of human Muscarinic Acetylcholine Receptor M2/CHRM2 (P08172). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?