missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Muscarinic Acetylcholine Receptor M1/CHRM1 Overexpression Lysate
Shop All Bio Techne ProductsDescription
The Muscarinic Acetylcholine Receptor M1/CHRM1 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
| Cell Line | The lysate was created in HEK293T cells, using plasmid ID RC25794 and based on accession number NM_738. The protein contains a C-terminal DDK tag. |
| For Use With (Application) | Western Blot |
| Quantity | 0.1 mg |
| Storage Buffer | RIPA buffer |
| Purity | Protein |
| Test Specificity | Homo sapiens cholinergic receptor, muscarinic 1 (CHRM1), mRNA. |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Research Category | Cell Cycle and Replication, GPCR |
| Protein State | Native |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?