missing translation for 'onlineSavingsMsg'
Learn More

Musashi-2 Antibody - BSA Free, Novus Biologicals™

Product Code. 18602749 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.03mg
0.1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18602749 0.1 mg 0.1mg
18672849 0.025 mg 0.03mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18602749 Supplier Novus Biologicals Supplier No. NBP252978

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Musashi-2 Polyclonal antibody specifically detects Musashi-2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Musashi-2
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1.0 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 5 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS
Gene Accession No. B4DHE8
Gene Alias FLJ36569, MGC3245, MSI2H, musashi homolog 2 (Drosophila), musashi-2, RNA-binding protein Musashi homolog 2
Host Species Rabbit
Immunogen Full length recombinant human Mushashi 2 protein [UniProt B4DHE8]
Purification Method Immunogen affinity purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 124540
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.