Learn More
Invitrogen™ MUS81 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143846
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: rat brain tissue. Flow: 293T cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Interacts with EME1 and EME2 to form a DNA structure-specific endonuclease with substrate preference for branched DNA structures with a 5'-end at the branch nick. Typical substrates include 3'-flap structures, replication forks and nicked Holliday junctions. May be required in mitosis for the processing of stalled or collapsed replication forks.
Specifications
| MUS81 | |
| Polyclonal | |
| Unconjugated | |
| MUS81 | |
| 1200008A18Rik; AI182501; AW045863; crossover junction endonuclease MUS81; FLJ21012; FLJ44872 MUS81; MUS81; MUS81 endonuclease homolog; MUS81 endonuclease homolog (yeast); MUS81 structure-specific endonuclease subunit; SLX3; SLX3 structure-specific endonuclease subunit homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 293678, 80198 | |
| -20°C | |
| Lyophilized |
| ELISA, Flow Cytometry, Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| Q4KM32, Q96NY9 | |
| MUS81 | |
| E.coli-derived human MUS81 recombinant protein (Position: R6-C545). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.