missing translation for 'onlineSavingsMsg'
Learn More

Murine Hepatitis Virus A59 nsp9 Antibody (2C6.H1), DyLight 650, Novus Biologicals™

Product Code. 18409216 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409216 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18409216 Supplier Novus Biologicals Supplier No. NBP221671C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Murine Hepatitis Virus A59 nsp9 Monoclonal antibody specifically detects Murine Hepatitis Virus A59 nsp9 in Virus samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Dot Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Murine Hepatitis Virus A59 nsp9
Applications Western Blot, Immunocytochemistry/Immunofluorescence, Dot Blot
Classification Monoclonal
Clone 2C6.H1
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias 2'-O-MT, 3' to 5' exonuclease, 3C-like proteinase, HEL1, leader protein, NendoU, nsp10, nsp11, NSP2, nsp4, nsp6, nsp7, nsp8, nsp9, ORF1a polyprotein, ORF1ab polyprotein, PLpro, polyprotein pp1a, polyprotein pp1ab, RNA-dependent RNA polymerase
Host Species Mouse
Immunogen This Murine Hepatitis Virus A59 nsp9 Antibody (2C6.H1) was produced in mice by repeated immunizations with E.coli derived full-length Murine Hepatitis Virus A59 nsp9 protein. This protein is part of the viral replicase polyprotein. (Uniprot: P0C6V0)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1489749
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.