missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MURF3 Antibody (OTI7F8), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72827PE
This item is not returnable.
View return policy
Description
MURF3 Monoclonal antibody specifically detects MURF3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| MURF3 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI7F8 | |
| Western Blot, Immunohistochemistry | |
| MuRF, MuRF3, MURF-3, MURFtripartite motif-containing protein 54, Muscle-specific RING finger protein, Muscle-specific RING finger protein 3, RING finger protein 30muscle-specific RING-finger protein 3, RNF30, tripartite motif containing 54, tripartite motif-containing 54 | |
| Human recombinant protein fragment corresponding to amino acids 166-400 of human TRIM54 (NP_115935) produced in E.coli. | |
| 0.1 mL | |
| Zinc Finger | |
| 57159 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction