missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MURF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562826
This item is not returnable.
View return policy
Description
Immunogen sequence: SNDRVQGVIS QLEDTCKTIE ECCRKQKQEL CEKFDYLYGI LEERKNEMTQ VITRTQEEKL EHVRALIK Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 90%.
The protein encoded by this gene contains a RING zinc finger, a motif known to be involved in protein-protein interactions. This protein associates transiently with microtubules, myosin, and titin during muscle sarcomere assembly. It may act as a transient adaptor and plays a regulatory role in the assembly of sarcomeres. Four alternatively spliced transcript variants encoding distinct isoforms have been described.
Specifications
| MURF2 | |
| Polyclonal | |
| Unconjugated | |
| TRIM55 | |
| D830041C10Rik; MURF2; MuRF-2; muscle specific ring finger 2; muscle-specific RING finger protein 2; RING finger protein 29; RNF29; Trim55; tripartite motif containing 55; tripartite motif-containing 55; tripartite motif-containing protein 55 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84675 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BYV6 | |
| TRIM55 | |
| Recombinant protein corresponding to Human MURF2. Recombinant protein control fragment (Product #RP-93811). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction