missing translation for 'onlineSavingsMsg'
Learn More

MuRF1/TRIM63 Antibody [DyLight 405], Novus Biologicals™

Product Code. 30733986 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30733986 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733986 Supplier Novus Biologicals Supplier No. AF5366E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

MuRF1/TRIM63 Polyclonal antibody specifically detects MuRF1/TRIM63 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MuRF1/TRIM63
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias EC 6.3.2.-, IRFMURF2, Iris RING finger protein, MuRF1, MuRF-1, muscle specific ring finger protein 2, Muscle-specific RING finger protein 1, RING finger protein 28FLJ32380, RNF28E3 ubiquitin-protein ligase TRIM63, SMRZMURF-1, Striated muscle RING zinc finger protein, tripartite motif containing 63, tripartite motif-containing 63, Tripartite motif-containing protein 63
Host Species Goat
Immunogen E. coli-derived recombinant human MuRF1/TRIM63, Met1-Gly325, Accession # Q969Q1
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 84676
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.