missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MURF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596226
This item is not returnable.
View return policy
Description
Positive Samples: Mouse heart; Cellular Location: Cytoplasm, M line, Nucleus, Z line, myofibril, sarcomere Immunogen sequence: LDKSTKLVET AIQSLDEPGG ATFLLTAKQL IKSIVEASKG CQLGKTEQGF ENMDFFTLDL EHIADALRAI DFGTDEEEEE FIEEEDQEEE ESTEGKEEGH Q.
This gene encodes a member of the RING zinc finger protein family found in striated muscle and iris. The product of this gene is localized to the Z-line and M-line lattices of myofibrils, where titin's N-terminal and C-terminal regions respectively bind to the sarcomere. In vitro binding studies have shown that this protein also binds directly to titin near the region of titin containing kinase activity. Another member of this protein family binds to microtubules. Since these family members can form heterodimers, this suggests that these proteins may serve as a link between titin kinase and microtubule-dependent signal pathways in muscle.
Specifications
| MURF1 | |
| Polyclonal | |
| Unconjugated | |
| TRIM63 | |
| E3 ubiquitin-protein ligase TRIM63; FLJ32380; IRF; iris ring finger protein; Murf; MURF 1; MuRF1; MuRF-1; MURF2; muscle ring finger protein 1; muscle specific ring finger protein 2; muscle-specific RING finger protein 1; OTTHUMP00000008701; RF1; ring finger protein 28; RING-type E3 ubiquitin transferase TRIM63; RNF 28; RNF28; SMRZ; striated muscle RING zinc finger protein; TRIM 63; Trim63; tripartite motif containing 63; tripartite motif containing 63, E3 ubiquitin protein ligase; tripartite motif protein 63; tripartite motif-containing 63; tripartite motif-containing protein 63 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 140939, 433766, 84676 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.99 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q38HM4, Q91Z63, Q969Q1 | |
| TRIM63 | |
| Synthetic peptide | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction