missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MURC Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA525555
This item is not returnable.
View return policy
Description
MURC induces RHOA activation and activates NPPA transcription and myofibrillar organization through the Rho/ROCK signaling pathway.
Specifications
| MURC | |
| Polyclonal | |
| PBS with 0.09% sodium azide | |
| A2AMM0, Q5BKX8 | |
| CAVIN4 | |
| KLH conjugated synthetic peptide between 87-116 amino acids from the N-terminal region of human MURC | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| CAVIN4 | |
| 2310039E09Rik; Caveolae-associated protein 4; cavin 4; CAVIN4; cavin-4; Murc; muscle related coiled-coil protein; muscle-related coiled-coil protein; Muscle-restricted coiled-coil protein | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 347273, 68016 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction