missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MURC Polyclonal Antibody
GREENER_CHOICE

Product Code. 15732231 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15732231 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15732231 Supplier Invitrogen™ Supplier No. PA554296

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MEHNGSASNA DKIHQNRLSS VTEDEDQDAA LTIVTVLDKV ASIVDSVQAS QKRIEERHRE MENAIKSVQI DLLKLSQSHS NTGHIINKLF Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.

MURC induces RHOA activation and activates NPPA transcription and myofibrillar organization through the Rho/ROCK signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MURC
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CAVIN4
Gene Accession No. Q5BKX8
Gene Alias 2310039E09Rik; Caveolae-associated protein 4; cavin 4; CAVIN4; cavin-4; Murc; muscle related coiled-coil protein; muscle-related coiled-coil protein; Muscle-restricted coiled-coil protein
Gene Symbols CAVIN4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MURC. Recombinant protein control fragment (Product #RP-93060).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 347273
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.