missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MUPP1 Polyclonal antibody specifically detects MUPP1 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | MUPP1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 405 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | DKFZp781P216, FLJ25909, FLJ90240, Multi-PDZ domain protein 1, multiple PDZ domain protein, MUPP1FLJ34626 |
| Host Species | Rabbit |
| Immunogen | The immunogen for this product maps to a region between residue 1125 and 1150 of human Multiple PDZ Domain Protein using the numbering given in entry NP_003820.2 (GeneID 8777). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?