missing translation for 'onlineSavingsMsg'
Learn More

Mupirocin, MedChemExpress

Product Code. 16498638
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16498638

Brand: Medchem Express HYB0958

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mupirocin (BRL-4910A, Pseudomonic acid) is an orally active antibiotic isolated from Pseudomonas fluorescens. Mupirocin apparently exerts its antimicrobial activity by reversibly inhibiting isoleucyl-transfer RNA, thereby inhibiting bacterial protein and RNA synthesis.

Mupirocin (BRL-4910A, Pseudomonic acid) is an orally active antibiotic isolated from Pseudomonas fluorescens. Mupirocin apparently exerts its antimicrobial activity by reversibly inhibiting isoleucyl-transfer RNA, thereby inhibiting bacterial protein and RNA synthesis.

Specifications

Chemical Name or Material Mupirocin
Physical Form Solid
CAS 12650-69-0
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C26H44O9
For Use With (Application) COVID-19-immunoregulation
Synonym BRL-4910A Pseudomonic acid
Solubility Information DMSO : ≥ 100 mg/mL (199.75 mM)
SMILES O[C@@H]1[C@@](CO[C@@H](C/C(C)=C/C(OCCCCCCCCC(O)=O)=O)[C@@H]1O)([H])C[C@H]2[C@@]([C@@H](C)[C@@H](O)C)([H])O2
Molecular Weight (g/mol) 500.62
Formula Weight 500.62
Percent Purity 98.34%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.