missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mupadolimab Recombinant Monoclonal Antibody

Recombinant Monoclonal Antibody
£397.00 - £1002.00
Specifications
| Antigen | Mupadolimab Humanized |
|---|---|
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30282479
![]() |
Invitrogen™
MA559501 |
100 μg |
£397.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30282756
|
Invitrogen™
MA559502 |
1 mg |
£1002.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Mupadolimab is a humanized monoclonal antibody that targets CD73, an enzyme expressed on various immune cells, including B cells and T cells. CD73 plays crucial roles in processes such as immunomodulation and adenosine production. CD73 (also known as ecto-5'-nucleotidase) is encoded by the NT5E gene located on chromosome 6q14-q21. It is a glycosylphosphatidylinositol (GPI)-anchored membrane protein that catalyzes the dephosphorylation of extracellular AMP to adenosine and inorganic phosphate. Functionally, CD73 plays critical roles in various physiological and pathological processes. It modulates immune response by generating adenosine, which has immunosuppressive effects, thus aiding tumor cells in evading the immune system. CD73 is implicated in cancer cell proliferation, migration, invasion, and tumor angiogenesis, making it a significant target for cancer immunotherapy. Its expression is also associated with memory T-cell function, suggesting its pivotal role in adaptive immunity. Moreover, mutations in NT5E are linked to a rare genetic disorder known as arterial calcifications due to deficiency of CD73.Specifications
| Mupadolimab Humanized | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Lyophilized | |
| IgG1 | |
| Human | |
| Primary | |
| Protein A |
| 1 mg/mL | |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| Unconjugated | |
| Human | |
| RUO | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title