missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MUNC18 Recombinant Rabbit Monoclonal Antibody (JE38-54)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556520
This item is not returnable.
View return policy
Description
MUNC18 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
This gene encodes a syntaxin-binding protein. The encoded protein appears to play a role in release of neurotransmitters via regulation of syntaxin, a transmembrane attachment protein receptor. Mutations in this gene have been associated with infantile epileptic encephalopathy-4. Alternatively spliced transcript variants have been described.
Specifications
| MUNC18 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O08599, P61764, P61765 | |
| Stxbp1 | |
| Recombinant protein within human munc18-1 aa 401-500/594. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| JE38-54 | |
| Unconjugated | |
| Stxbp1 | |
| AI317162; AI326233; ANC18HA; EIEE4; FLJ37475; hUNC18; MGC146482; minisatellite 10g detected by probe MMS10; MMS10-G; Ms10g; MUNC18-1; Munc-18a; munc-18c; neuronal SEC1; NSEC1; n-sec1; NSEC1A; p67; Protein unc-18 homolog 1; protein unc-18 homolog A; rbSec1; Rb-sec1; rbSec1A; rbSec1B; rop; RP11-56D16.3; Sec1; Stxbp1; Sxtbp1; syntaxin binding protein 1; syntaxin-binding protein 1; UNC18; UNC-18 homolog; unc-18-1; Unc18-1; UNC18A; unc-18A; Unc18h | |
| Rabbit | |
| Protein A | |
| RUO | |
| 20910, 25558, 6812 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction