missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUM1 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00084939-B01P
This item is not returnable.
View return policy
Description
MUM1 Polyclonal antibody specifically detects MUM1 in Human samples. It is validated for Western Blot
Specifications
| MUM1 | |
| Polyclonal | |
| Western Blot 1:500 | |
| ENSP00000345789 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| EXPAND1, FLJ14868, FLJ22283, melanoma associated antigen (mutated) 1, melanoma ubiquitous mutated protein, MGC131891, MGC163315, MUM-1HSPC211, Mutated melanoma-associated antigen 1, Protein expandere, PWWP domain-containing protein MUM1 | |
| MUM1 (ENSP00000345789, 1 a.a. - 711 a.a.) full-length human protein. MMADAKYVLCRWEKRLWPAKVLARTATSTKNKRRKEYFLAVQILSLEEKIKVKSTEVEILEKSQIEAIASSLASQNEVPAAPLEELAYRRSLRVALDVLSEGSIWSQESSAGTGRADRSLRGKPMEHVSSPCDSNSSSLPRGDVLGSSRPHRRRPCVQQSLSSSFTCEKDPECKVDHKKGLRKSENPRGPLVLPAGGGAQDESGSRIHHKNWTLASKRGGNSAQKASLCLNGSSLSEDDTERDMGSKGGSWAAPSLPSGVREDDPCANAEGHDPGLPLGSLTAPPAPEPSACSEPGECPAKKRPRLDGSQRPPAVQLEPMAAGAAPSPGPGPGPRESVTPRSTARLGPPPSHASADATRCLPCPDSQKLEKECQSSEESMGSNSMRSILEEDEEDEEPPRVLLYHEPRSFEVGMLVWHKHKKYPFWPAVVKSVRQRDKKASVLYIEGHMNPKMKGFTVSLKSLKHFDCKEKQTLLNQAREDFNQDIGWCVSLITDYRVRLGCGSFAGSFLEYYAADISYPVRKSIQQDVLGTKLPQLSKGSPEEPVVGCPLGQRQPCRKMLPDRSRAARDRANQKLVEYIVKAKGAESHLRAILKSRKPSRWLQTFLSSSQYVTCVETYLEDEGQLDLVVKYLQGVYQEVGAKVLQRTNGDRIRFILDVLLPEAIICAISAVDEVDYKTAEEKYIKGPSLSYREKEIFDNQLLEERNRRRR | |
| 0.05 mg | |
| Melanoma Cell Markers | |
| 84939 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction