missing translation for 'onlineSavingsMsg'
Learn More

Ansell Edmont™ Multitop 40 Immersion Gloves

Product Code. 15617473 Shop All Ansell Products
Click to view available options
Size:
8
9
10
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 15617473

Brand: Ansell Edmont™ A192100

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Colored over-dip acts as a wear indicator. Ansell Edmont™ Multitop 40 Immersion Gloves have been coated by a multilayer process to provide exceptional mechanical and chemical protection in industrial and chemical applications.

  • Coated by multilayer process:
    • Grey colored PVC integral base coating for mechanical protection
    • Superior orange colored over-dip coating covering palm and fingers for chemical protection
  • Mechanical finish by microgranulation
  • Excellent resistance to abrasion
  • Excellent chemical resistance to acids, alkalis, cutting oils, petrol and gasoline
TRUSTED_SUSTAINABILITY

Specifications

Back Material PVC
Certifications/Compliance European Directive 89/686/EEC, EN 420:2003, EN388:2003, EN374:2003
Classification Category III
Coating Material PVC
Color Gray
Exterior Finish Microgranulated
Size 10
Sterility Non-sterile
Glove Cuff Style Gauntlet
Liner Material Interlock cotton
Packaging Type Bagged
Puncture Rating AQL 0.65
Quantity 12 Pair
Resistant To Acids, alkalis, cutting oils, petrol and gasoline
Length (Metric) 400 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.