missing translation for 'onlineSavingsMsg'
Learn More

ZVG™ Multitex™ z47 Cleaning Cloth Roll

Product Code. 17803535
Change view
Click to view available options
Color:
White
Unit Size:
1 roll
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
17803535 White 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17803535 Supplier ZVG™ Supplier No. 1114700
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

This high-quality cleaning cloth roll made from viscose nonwoven material is very soft and highly tear-resistant, for various cleaning tasks.

The Multitex™ cloths can be washed and wrung out during cleaning. They are regularly used in sensitive work areas such as precision mechanics, paint shops, the electrical and automotive industries, as well as in nursing homes. The Multitex™ z70 cloth stands out due to its increased material with higher absorption capacity. This allows for more liquid to be absorbed without tearing the material.

  • Viscose nonwoven fabric with textile character
  • Suitable for both wet and dry cleaning
  • Ideal for degreasing
  • Very thick wipes (medium strength, between z60 and z80)
  • Very soft, yet lint-free
  • Tear- and solvent-resistant
  • Silicone-free
  • Each roll contains 475 tear-off sheets

Viscose fibers are industrially produced from the raw material cellulose. The chemical composition of this 'natural synthetic fiber' is similar to that of cotton. Viscose products are absorbent, flexible, and durable due to their textile character.

Specifications

Product Type Cleaning Cloth
Length (English) 15.7 in.
Length (Metric) 40 cm
Width (English) 11.8 in.
Width (Metric) 30 cm
Material Polypropylene, Viscose, Cotton
Color White
Disposable Single-use
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.