missing translation for 'onlineSavingsMsg'
Learn More

BD Biosciences™ Multitest CD3 FITC/CD16 + CD56 PE/CD45 PerCP/CD19 APC with BD Trucount™ Tubes (CE/IVD)

Four-color direct immunofluorescence reagent for identifying and determining the percentages of human T, natural killer, and B lymphyocytes in erhythrocyte-lysed whole blood.

Brand:  BD Biosciences™ 342446

Additional Details : Weight : 0.44600kg

Product Code. 15836008

  • £2230.00 / 1 set
Estimated Shipment: 14-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Clone SJ25C1 (also known as SJ25-C1) SK7 (also known as Leu-4) NCAM16.2 (also known as NCAM 16) B73.1 2D1 (CE/IVD).
  • BD MultiTEST™ CD3 fluorescein isothiocyanate (FITC)/CD16 + CD56 phycoerythrin† (PE)/CD45 peridinin chlorophyll protein (PerCP)/ CD19 allophycocyanin (APC) is a four-color direct immunofluorescence reagent for use with a suitably equipped flow cytometer to identify and determine the percentages and absolute counts of mature human T (CD3+), natural killer (NK) (CD3CD16+CD56+), and B (CD3CD19+) lymphocytes in erythrocytelysed whole blood.
  • When used with TruCOUNT™ Tubes, absolute counts of these populations can be enumerated from a single tube.
  • BD TriTEST reagent and TruCOUNT Tubes can be used with the FACS Loader.
  • Contents:
  • Specificity: CD19: SJ25C1 (SJ25-C1), Format: APC, Isotype: IgG<sub>1</sub> K, Entrez Gene ID: 930
  • Specificity: CD3, Clone: SK7(Leu-4), Format FITC, Isotype IgG<sub>1</sub> K, Entrez Gene ID: 916
  • Specificity: CD56 (NCAM-1), Clone: NCAM16.2 (NCAM 16), Format: PE, Isotype: IgG<sub>2b</sub> K, Entrez Gene ID: 4684
  • Specificity: CD16 (FcγRIII), Clone: B73.1, Format: PE, Isotype: IgG<sub>1</sub> K
  • Specificity: CD45, Clone: 2D1, Format: PerCP, Isotype: IgG<sub>1</sub> K
Specifications

Specifications

Trucount™ Tubes
50 Tests
CE/IVD
Store the reagent at 2°C–8°C to maintain stability. Do not use after expiration date shown on the label. Do not freeze the reagent or expose it to direct light during storage or during incubation with cells. Keep the reagent vial dry. Do not use the reagent if you observe any changes in appearance. Precipitation or discoloration indicates instability or deterioration.
Immunofluorescence Flow Cytometry Test Reagents
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers