Learn More
MP Biomedicals MULTISURE™ HIV 1/2 Confirmatory Test
Product Code. 31605226 Shop All MP Biomedicals Products
Change view
Click to view available options
Detectable Analytes:
HIV-1 gp120, HIV-1 gp41, HIV-1 p24, HIV-2 gp36, HIV-2 gp105
Unit Size:
20 tests
Product Code. Detectable Analytes Unit Size
31605226 HIV-1 gp120, HIV-1 gp41, HIV-1 p24, HIV-2 gp36, HIV-2 gp105 20 tests
1 options
Product Code. 31605226 Supplier MP Biomedicals Supplier No. 93030020
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Qualitative rapid in vitro immunochromatographic assay to detect and differentiate HIV-1 and HIV-2 antibodies in serum or plasma
  • MP Diagnostics MULTISURE™ HIV 1/2 Confirmatory Test is intended as a rapid confirmatory test, developed to detect and differentiate antibodies specific to HIV-1 gp120, HIV-1 gp41, HIV-1 p24 and HIV-2 gp36, HIV-2 gp105 antigens in human serum or plasma.
  • Confirm and Differentiate HIV-1 and HIV-2
  • Accurate Result
  • Simple Workflow
  • Flexible: Manual or Automatic Reading of Result
  • Wide Temperature Storage (2 °C to 30 °C)
Specifications
CE Marker CE-IVD
Detectable Analytes HIV-1 gp120, HIV-1 gp41, HIV-1 p24, HIV-2 gp36, HIV-2 gp105
Type HIV 1/2 Confirmatory Test