missing translation for 'onlineSavingsMsg'
Learn More

ZVG™ Multisoft™ Air Wipes

Product Code. 17893545
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17893545 White Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17893545 Supplier ZVG™ Supplier No. 1010203
In Stock
Add to basket
Add to basket

These Air Wipes are made of Airlaid material for demanding cleaning applications and are lint-free and highly absorbent. Ideal for sensitive surfaces.

Sensitive surfaces require gentle treatment. These extra soft, low-lint specialty cloths are an ideal choice for care and polishing tasks. They efficiently absorb liquids such as polishing wax and evenly release it without compromising their strength.

  • Airlaid material
  • Embossed white cellulose
  • Extra soft
  • Latex- and silicone-free
  • Solvent-resistant
  • Suitable for sensitive surfaces

Specifications

Product Type Wipe
Length (English) 10.6 in.
Length (Metric) 27 cm
Width (English) 12.6 in.
Width (Metric) 32 cm
Material Fleece
Color White
Disposable Single-use
Packaging Quantity 1000 Towels
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.