missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MultiShot™ StripWell DH5α T1R Competent Cells

Product Code. 16378741
Change view
Click to view available options
Quantity:
1 x 96 tube rack (50 μl/tube)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16378741 1 x 96 tube rack (50 μl/tube) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16378741 Supplier Invitrogen™ Supplier No. C449601
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

Box 1:

  • MultiShot StripWell DH5α T1R Chemically Competent E. coli
  • One rack of 12 strips x 8-tubes, 50 μl per tube

Box 2:

  • pUC19 DNA (10 pg/μl): 1 vial, 50 μl
  • S.O.C. medium: 2 bottles, 15ml each

MultiShot StripWell DH5α T1R chemically competent E. coli cells are cloning competent cells packaged in a rack containing 12 strips of 8 tubes to increase productivity for medium-throughput bacterial transformations.

MultiShot StripWell DH5α T1R chemically competent E. coli cells are cloning competent cells packaged in a rack containing 12 strips of 8 tubes to increase productivity for medium-throughput bacterial transformations. This strain is derived from the popular DH5α strain, one of the most frequently used laboratory cloning strains. All of our T1R strains have the tonA (also known as fhuA) mutation in their genotype that confers resistance to T1 and T5 phages. A resistant genotype offers added security to protect valuable clones and libraries as T1 bacteriophage can rapidly spread among E. coli hosts and contaminate repositories.

DH5α T1R for routine cloning applications
The DH5α strain is well-known and broadly used laboratory strain for chemical transformations as it is ideal for transformations of cloned unmethylated DNA. To increase productivity for medium-throughput bacterial transformations, the cloning competent DH5α T1R E. coli cells are packaged in racks containing 12 strips of 8.

MultiShot StripWell DH5α T1R Competent Cells include:
hsdR for efficient transformation of unmethylated DNA from PCR amplifications
lacZΔM15 for blue/white color screening of recombinant clones
endA1 for cleaner preparations of DNA and better results in downstream applications due to the elimination of non-specific digestion by endonuclease I
recA1 for reduced occurrence of non-specific recombination in cloned DNA
tonA (fhuA) for T1 and T5 phage resistance 

MultiShot StripWell flexible format
Each StripWell of eight connected tubes is designed with the same single-tube, single-use features as our One Shot format tubes. This format allows all steps of the transformation protocol, up to plating, to take place in the same tube, saving time and reducing chance of contamination. After addition of DNA, MultiShot StripWell competent cells can be transformed by heat-shock at 42°C in a water bath without transfering to a new tube.

The StripWell tubes can also be cut to permit single transformations. This feature eliminates wasted aliquots and avoids needless freeze-thaw cycles that can result in reduced transformation efficiency. 

Benifits of the MultiShot StripWell format include:
• Increased productivity—suitable for high throughput experimentation 
• Maximal flexibility—transform as few as 1 tube or as many as 96 from the StripWell rack
• Familiar convenience—add DNA directly to cells, heat shock, and recover in the same tube

Genotype
F- φ80lacZΔM15 Δ(lacZYA-argF)U169 recA1 endA1 hsdR17 (rK–, mK+) phoA supE44 λ thi-1 gyrA96 relA1 tonA

Find the strain and format that you need
• We offer a variety of DH strains in both chemically competent and electrocompetent cell formats.
• For high-throughput transformation options, choose from our collection of MultiShot Formatted Competent Cells.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemically Competent Cells
Contains F' Episome No
Cloning Methylated DNA No
Content And Storage • MultiShot StripWell DH5α T1R Competent Cells (1 x 96-well plate at 50 μL/well)
Store at –80°C.

• pUC19 DNA (50 μL at 10 pg/μL)
Store at –20°C.

• S.O.C. Medium (2 x 15 mL)
Store at 4°C or room temperature.

Antibiotic Resistance Bacterial No
Blue/White Screening Yes (lacZΔM15)
High-throughput Compatibility High
Preparing Unmethylated DNA No
Reduces Recombination Yes (recA1)
Shipping Condition Dry Ice
T1 Phage - Resistant (tonA) Yes
Species E. coli (K12)
Format 8-well Strips
Product Line Multishot
Quantity 1 x 96 tube rack (50 μl/tube)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.