missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MultiShot™ BL21(DE3) Chemically Competent E. coli Cells, 96-well plate

Product Code. 31781154
Change view
Click to view available options
Quantity:
1 x 96 Well Plate (20 μL/Well)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31781154 1 x 96 Well Plate (20 μL/Well) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31781154 Supplier Invitrogen™ Supplier No. A40007051
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MultiShot BL21(DE3) Chemically Competent E. coli Cells enable high-throughput protein expression with compatibility for automated liquid handling. This ready-to-use format streamlines cloning and expression workflows using the reliable BL21(DE3) expression strain.

Invitrogen MultiShot BL21(DE3) Chemically Competent E. coli Cells are suitable for the expression of non-toxic heterologous genes. They are pre-aliquoted into a 96-well PCR plate to increase transformation productivity.

Features of MultiShot BL21(DE3) competent cells include:

  • High transformation efficiency: >1x107 cfu/μg pUC19 DNA
  • Specifically constructed for high-level expression of recombinant proteins
  • Microtiter plate format design convenient for automated high-throughput cloning
  • Competent cells pre-aliquoted into 96-well plate, ready for transformation
  • S.O.C. Outgrowth medium included

MultiShot BL21(DE3) competent cells in a 96-well plate are designed to facilitate high-throughput transformation compatible with liquid handling equipment. The BL21 (DE3) strain contains the lambda DE3 prophage that carries the gene for T7 RNA polymerase under control of a lacUV5 promoter, allowing expression of the T7 RNA polymerase to be induced with IPTG. BL21(DE3) is an E. coli B strain and does not contain the lon protease. It is also deficient in the outer membrane protease OmpT. The lack of these two key proteases reduces degradation of heterologous proteins expressed in the cells.

Genotype

F– ompT hsdSB (rB–, mB–) gal dcm (DE3)

TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemically competent cells
Includes pUC19 DNA, S.O.C media
Transformation Efficiency Level ≥1.0 x 107
Shipping Condition Dry Ice
Species E. coli
Format 96-well Plate
Product Line MultiShot
For Use With (Application) Protein Expression
Quantity 1 x 96 Well Plate (20 μL/Well)

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.