missing translation for 'onlineSavingsMsg'
Learn More

Merck MultiScreenHTS Durapore™ 96-Well Filter Plates

Product Code. 10011953 Shop All Merck Products
Change view
Click to view available options
Pore Size:
0.22 μm
0.45 μm
0.65 μm
1.2 μm
Color:
Clear
Opaque
Material:
Barex
Styrene
Quantity:
10/Pk.
50/Pk.
Unit Size:
Pack of 10
Pack of 50
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Color Material Quantity unitSize
10011953 1.2 μm Opaque Barex 50/Pk. Pack of 50
10659113 1.2 μm Clear Styrene 50/Pk. Pack of 50
10777264 1.2 μm Clear Styrene 10/Pk. Pack of 10
10432753 1.2 μm Clear Styrene 10/Pk. Pack of 10
10400334 0.65 μm Opaque Barex 50/Pk. Pack of 50
10367123 0.22 μm Clear Styrene 10/Pk. Pack of 10
10154213 0.22 μm Clear Styrene 50/Pk. Pack of 50
10347133 0.22 μm Opaque Barex 50/Pk. Pack of 50
10715164 0.45 μm Opaque Barex 10/Pk. Pack of 10
10367513 0.45 μm Clear Styrene 10/Pk. Pack of 10
10408993 0.45 μm Clear Barex 50/Pk. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 10011953 Supplier Merck Supplier No. MSBVN1B50
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Designed for full automation compatibility. MultiScreenHTS Durapore™ 96-Well Filter Plates are innovative plates ideal for bead based assays. Opaque Barex™ and clear, sterile or non-sterile styrene and racks are available in a range of pore sizes.

For use in receptor-ligand binding, general sample preparation, ELISPOT, enzyme activity, bead-based assays, whole organism visualization, and radioisotope detection.
  • Standardized dimensions and aligned, rigid sidewalls
    • Allows full compatibility with automated/robotic gripper arms, stackers, plate washers, bar code labels, and plate readers
  • Unique two-piece plate design eliminates surface contact with individual well “drip directors” to prevent crosstalk between wells
  • Extended skirt and vacuum manifolds
    • Allows easy, contamination-free stacking directly with collection plates
    • Protects recessed underdrain
    • Provides ample surface for bar code labels
    • Improves vacuum seal
  • Removable underdrain allows membrane access for ELISPOT assays, whole organism visualization, and radioisotope detection
  • Isolated well design and individually sealed filter discs eliminate potential for reagent flow from well to well
  • Opaque plastic prevents light transfer between wells during direct scintillation counting
  • Low binding hydrophilic Durapore™ membrane allows low nonspecific binding of antibodies, reagents, and samples
  • Meet ANSI™/SBS 2004 plate specifications
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI, SBS
Format Standard
Membrane Hydrophilic PVDF
Array 8 x 12
Pore Size 1.2 μm
Color Opaque
Material Barex
No. of Wells 96
Sterility Non-sterile
Filtration Area 0.28 cm2
For Use With (Application) Bead based assays
Quantity 50/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.