missing translation for 'onlineSavingsMsg'
Learn More

Merck MultiScreenHTS 384-Well Filter Plates

Product Code. 10500275 Shop All Merck Products
Change view
Click to view available options
Pore Size:
1 μm
1.2 μm
Quantity:
10/Pk.
50/Pk.
Unit Size:
Pack of 10
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Quantity unitSize
10500275 1.2 μm 10/Pk. Pack of 10
11542144 1 μm 50/Pk. Pack of 50
10602605 1 μm 10/Pk. Pack of 10
11784694 1.2 μm 50/Pk. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10500275 Supplier Merck Supplier No. MZFCN0W10
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use in automated, high throughput applications, including biological assays and sample preparation. MultiScreenHTS 384-Well Filter Plates are non-sterile, single use plates available in several mesh materials and pore sizes to meet the needs of different applications.

For use in receptor-ligand binding, general sample preparation, enzyme assays with precipitation and DNA purification.
  • Rigid sidewalls allows full compatibility with automated/robotic systems
  • Unique plate design and individually sealed filter discs prevent cross-talk between wells
  • Opaque plastic lids prevents light transfer between wells during direct scintillation counting
  • Barcoding available
  • Meet ANSI™/SBS standards
TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Membrane Glass Fiber, Polyester Mesh
Array 16 x 24
Pore Size 1.2 μm
Color Opaque, White Bottom
Product Type MultiScreenHTS-FC Plate
Material Barex
No. of Wells 384
Sterility Non-sterile
Filtration Area 0.09cm2
For Use With (Application) Receptor, ligand binding assays, sample prep, TCA precipitation
Quantity 10/Pk.
Dimensions (L x W x H) 1.28 x 8.55 x 1.44 cm
Length (Metric) 1.28 cm
Width (Metric) 8.55 cm
Height (Metric) 1.44 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.