Learn More
Merck MultiScreen Punch Carrier
Product Code. 10186720 Shop All Merck Products
Change view
Click to view available options
Quantity:
1Each
Unit Size:
Each
Product Code. Quantity unitSize
10186720 1Each Each
1 options
Product Code. 10186720 Supplier Merck Supplier No. MACP09600
Only null left
Add to basket
Add to basket
Order Info
dimForUseWithApplication
TRUSTED_SUSTAINABILITY
Specifications
Product Type MultiScreen Punch Carrier
Quantity 1Each
For Use With (Application) This multiwell plate punch carrier is used for Radionucleotide counting by transfer of individual filters to tubes