missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Fully Compatible with Automation
High throughput MultiScreen™&subscript_begin;HTS&subscript_end; filter plates are specifically developed for use with automation. The 96 well and 384 well plates are designed to standardised dimensions and meet ANSI/SBS 2004 plate specifications. They are fully compatible with automated gripper arms, stackers, barcode labels, and plate readers.
Innovative Plate Design
Rigid sidewalls are aligned for easy handling by robotic gripper arm and also provide ample surfaces for bar code labels. The plate design protects the underdrain and eliminates surface contact with individual well 'drip directors'. The plastic skirt enables stacking directly with collection plates and improves the vacuum seal when used with a Millipore™ vacuum manifold. The removable underdrain gives full access to membranes and is ideal for applications including Elispot, radioisotope detection and whole organism visualisation.
Extensive Product Range
MultiScreen™&subscript_begin;HTS&subscript_end; filter plates are available in a broad range of Millipore plastics and membranes. MultiScreen™ Solubility filter plates are also optimised in HTS format for filtration-based solubility determination. Accessory column loaders are available for loading chromatography media for column separators. MultiScreen™&subscript_begin;HTS&subscript_end; filter plates are available in clear, opaque and white. For non-standard plastic and membrane combinations, custom requests are accepted. Available membranes include:
High throughput MultiScreen™&subscript_begin;HTS&subscript_end; filter plates are specifically developed for use with automation. The 96 well and 384 well plates are designed to standardised dimensions and meet ANSI/SBS 2004 plate specifications. They are fully compatible with automated gripper arms, stackers, barcode labels, and plate readers.
Innovative Plate Design
Rigid sidewalls are aligned for easy handling by robotic gripper arm and also provide ample surfaces for bar code labels. The plate design protects the underdrain and eliminates surface contact with individual well 'drip directors'. The plastic skirt enables stacking directly with collection plates and improves the vacuum seal when used with a Millipore™ vacuum manifold. The removable underdrain gives full access to membranes and is ideal for applications including Elispot, radioisotope detection and whole organism visualisation.
Extensive Product Range
MultiScreen™&subscript_begin;HTS&subscript_end; filter plates are available in a broad range of Millipore plastics and membranes. MultiScreen™ Solubility filter plates are also optimised in HTS format for filtration-based solubility determination. Accessory column loaders are available for loading chromatography media for column separators. MultiScreen™&subscript_begin;HTS&subscript_end; filter plates are available in clear, opaque and white. For non-standard plastic and membrane combinations, custom requests are accepted. Available membranes include:
- Hydrophilic Durapore™ PVDF membrane
- Glass fibre
- Hydrophobic Immobilon™-P membrane
- HA mixed cellulose esters
- Negatively charged phosphocellulose
- Positively charged DEAE
- Track-etched polycarbonate Applications include: receptor-ligand binding; general sample preparation; Elispot; enzyme activity; bead-based assays; whole organism visualisation and radioisotope detection.
Specifications
Specifications
| Format | Standard |
| Array | 8 x 12 |
| Material | Polystyrene |
| No. of Wells | 96 |
| For Use With (Application) | Collection plate for 96-well plates |
| Skirt Color | Clear |
| Well Color | Clear |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?