Learn More
Axygen™ MultiRack Tip Station
Product Code. 16312227 Shop All Axygen Scientific Products
Click to view available options
Volume (Metric):
10 μL
200 μL
Rack Color:
Red
Yellow
Sterility:
Non-sterile
Sterile
Tip Style:
Extended Length
Universal
Unit Size:
Pack of 960
Product Code. 16312227
Brand: Axygen™ MR10XTTS
Only null left
Add to basket
Edge
Add to basket
MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an interchangeable complete system offered in Bulk, Racked, Filter and Reload packaging styles to meet the variety of needs in the lab setting.
The Axygen MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an interchangeable complete system offered in bulk, racked, filter, and reload packaging styles to meet the variety of needs in the lab setting. The MultiRack tip system offers standard and extended length tips covering volumes from 1 μL to 1250 μL. For racked and reload packaging styles, the tips are pre-loaded into easy-load color-coded deck inserts. With the smaller footprint and reusable rack, this system uses 30% less plastic, minimizing the environmental impact. The stackable design saves critical bench space. Axygen MultiRack Tip System products are made in the USA and meet ISO standards. All tips are free of detectable RNase, DNase, DNA, and pyrogens. For more details visit our Certificates page at www.corning.com/lifesciences.
Features
• Reusable rack with reload systems 
• Smaller footprint and stackable design, saving valuable bench space
• Extended tips offered in 10, 300, and 1250 μL (1000 μL extended length) volumes
• Color-coded racks for easy volume identification
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Non-sterile
Color Red
Rack Color Red
Autoclavable Autoclavable
Certifications/Compliance ISO 13845
Clean Claims Free of detectable DNAse/RNAse, human DNA, and PCR inhibition
Format Individually Wrapped
Tip Style Extended Length
Packaging Tip Station
Quantity 192/layer, 5 layers/stack, 960/stack, 5 stacks/cs
Disposable Single-use
Graduated Yes
Product Line MultiRack
Material Polyethylene