missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Multiple Tumor Matched Tumor and Normal Tissue MicroArray
Shop All Bio Techne Products
Click to view available options
:
5 Slides
Specifications
Specifications
| Quantity | 5 Slides |
| Species | Human |
| For Use With (Application) | Please bake slides at 60C for 30 minutes before use. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction