missing translation for 'onlineSavingsMsg'
Learn More

Scientific Industries MultiPlate Genie

Product Code. 17886137
Click to view available options
Plug Type:
No Plug
UK Plug
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17886137

Brand: Scientific Industries SI4101

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

MultiPlate Genie is fully programmable microplate shaker for up to 8 plates providing consistent mxing in each well.

  • Digital high-speed vortex mixing microplate shaker handles up to 4 microplates, (or up to 8 plates with the optional accessory tray)
  • Combined speed and 1 mm orbit provides consistant well to well and plate to plate uniformity, even in 384-well plates
  • Featuring programable speed, time and continuous or pulse time, it’s ideal for achieving accurate and repeatable results
  • Pulse feature allows rapid pulsing or timed ''on/off'' mixing for interval mixing
  • Ideal for use in PCR Preparation, Digestions, Bead Mixing, ELISA, EIA, and Colorimetric Assay applications.

Specifications

Orbit 1 mm
Load Bearing Capacity (Metric) 499 g
Speed Range 500 to 2800 rpm
Electrical Requirements 230V, 50Hz
Voltage 230 V
Amperage 0.5 A
Rotation Orbital
Holds 4 Microplates
Product Type Shaker
Load Bearing Capacity (English) 1.1 lb.
Certifications/Compliance CE
Plug Type No Plug
Frequency 50 Hz
Depth (English) 15 in.
Depth (English) Tray 13.3 in.
Depth (Metric) 380 mm
Depth (Metric) Tray 338 mm
Height (English) 6 in.
Height (Metric) 150 mm
Length (English) 15 in.
Length (Metric) 380 mm
Width (English) 10 in.
Width (Metric) 250 mm
Width (Metric) Platform 241 mm
Shipping Weight (English) 14.5 lb.
Shipping Weight (Metric) 6.6 kg
Controller Type Digital Keyboard
Description MultiPlate Genie, 230V No Plug
Model MultiPlate Genie SI-4101
Display Digital Timer and Speed
For Use With (Application) PCR Preparation, Digestions, Bead Mixing, ELISA, EIA, and Colorimetric Assay applications
For Use With (Equipment) Microplates and Microplate Inserts
Humidity Range 0°C to 38°C (32°F to 100°F), 95% Humidity Max
Includes One tray for 4 microplates
No. of Positions 4
Timer Range 0 to 99 Minutes or Continuous
Warranty and Service Offering 24 months
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.