missing translation for 'onlineSavingsMsg'
Learn More

Kartell Multipet Pro 12 Channels

Product Code. 30048910
Click to view available options
Volume (Metric):
10 μL
20 μL
50 μL
100 μL
200 μL
300 μL
missing translation for 'unitSize'
Each
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 30048910

missing translation for 'mfr': Kartell 0174703

Solo quedan null
Añadir a la cesta
Añadir a la cesta
Este artículo no se puede devolver. Vea la política de devoluciones

Very precise, ergonomic light-weight design, with a structure made of polycarbonate, equipped with a large 4-digit display. Characterized by advanced technologies calibration and reduction of the dead space between the tip holder cone and the plunger. Fully autoclavable at 121 °C for 20 minutes.

  • Individually calibrated in an ISO 17025 accredited laboratory according to ISO 8655 standards. Calibration certificate is provided within the instrument.
  • Universal tip cones are compatible with most of the internationally accepted standard tips.
  • Patented UniCal™ technology facilitates a-lessthan-15 seconds quick recalibration in a single operation without disengaging the digits from the plunger mechanism and avoiding the traditional hit and trial method of calibration.
  • μAir™ technology ensures accuracy and precision better than the ISO 8655 standard, by minimizing dead air space between the piston and the liquid in the tip. This technology reduces the air compression and allow the user to achieve greater precision during laboratory practices.
  • Ultra Soft Plunger force, with its 1st step plunger force being only 8.95 N and the 2nd step plunger force being only 16.85 N. The soft plunger greatly improves user comfort even during long hours of pipetting.
  • Smooth outer surface, allowing easy cleaning.
  • Premium aesthetics and build quality, comparable with international top brands.
  • Ultra light weighted pipette (PC).
  • Polycarbonate housing material makes the micropipettes durable, lightweight, scratch proof and dirt resistant.
  • Spring and ball-based digit change mechanism ensures no accidental volume change while carrying out various operations.
  • The in-built tip ejector ejects tips with optimal force and provides great aesthetic value.
  • The different color coding of the dispensing button, facilitates the identification of the desired volume range during the daily laboratory routine
  • Useful for applications in Molecular Biology, Microbiology, Immunology, Analytical Chemistry, Research Laboratories, Educational Field etc.

Especificaciones

Color White
For Use With Standard Micropipette Tip
Number of Channels 12
Type Micropipette
Volume (Metric) 10 μL
Fit for Purpose Certified Quality
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado