missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Multifunctional Tool for M210 and BMP™21 Label Printers

Product Code. 17693215
Click to view available options
Color:
Black
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17693215

Brand: Brady™ 170424

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use this multifunctional tool to operate your label printer with one hand, leaving your other hand free.

The multifunctional accessory for Brady™ Label Printers consists of a magnet, flashlight, stand, belt loop and pole strap.

  • Ideal for an on-the-go labeling environment
  • Built in LED flashlight helps you create labels in low-lit areas
  • 2 AAA batteries required (not included)
  • Compatible with Brady™ BMP™21, BMP™21-LAB, M210, and M210-LAB Label Printers

Specifications

Color Black
For Use With (Equipment) BMP21 Label Printer, Brady M210 Label Printer
Includes Magnet, Flashlight, Stand, Belt Loop, Pole Strap
Certifications/Compliance 2011/65/EU and 2015/863
Alternate Part Number M210-TOOL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.