missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Multidrop™ Pico Digital Dispenser

Product Code. 16643823 Shop All Thermo Scientific Products
Change view
Click to view available options
Model:
Multidrop Pico 1 Digital Dispenser
Multidrop Pico 8 Digital Dispenser
Multidrop Pico 8 Surfactant Free Digital Dispenser
Dimensions (L x W x H):
38.5 x 28 x 19 cm (15 in x 11 in x 7.5 in)
47 x 38 x 23 cm (18.5 x 15 x 9.1 in)
47 x 38 x 23 cm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Dimensions (L x W x H) Unit Size
16643823 Multidrop Pico 8 Digital Dispenser 47 x 38 x 23 cm Each
31722884 Multidrop Pico 8 Surfactant Free Digital Dispenser 47 x 38 x 23 cm (18.5 x 15 x 9.1 in) Each
31337546 Multidrop Pico 8 Surfactant Free Digital Dispenser 47 x 38 x 23 cm (18.5 x 15 x 9.1 in) -
16671296 Multidrop Pico 1 Digital Dispenser 38.5 x 28 x 19 cm (15 in x 11 in x 7.5 in) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16643823 Supplier Thermo Scientific™ Supplier No. 5840600

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Multidrop™ Pico Digital Dispensers provide high-precision, non-contact dispensing from 8 pL to 10 μL, using up to ten times less reagent than manual pipetting. Ideal for miniaturizing assays, they ensure accurate results with user-friendly software.

The Multidrop™ Pico Digital Dispensers offer enhanced dispensing precision with volumes as low as 8 pL, using up to ten times less reagent than manual pipetting, reducing costs and waste. Designed for assay miniaturization, they boost productivity and improve accuracy, ideal for qPCR, dose response curves, drug screening assays, and enzyme profiling. The intuitive Multidrop™ PicoIT software supports complex experimental setups, enhancing productivity and data integrity while conserving resources and improving operational efficiency.

  • Cost Saving through assay miniaturization
  • Improved efficiency and sample integrity
  • Powerful, flexible, and easy to use software

Service Plan

Yes

TRUSTED_SUSTAINABILITY

Especificaciones

Product Type Digital Dispenser
Volume (Metric) 11 pL to 10 μL
For Use With (Application) Drug combination studies, Biochemical assays, Cell-based assays, DMPK, Secondary screening, Synthetic chemistry, assay development, qPCR, Enzyme profiling, Antibody therapies, SAR, and Spotting
Interface USB, PC Control
Model Multidrop Pico 8 Digital Dispenser
Plate Type 12, 24, 48, 96, 384, 1536 well plate, Plate height: 6-47 mm
Precision ≤ 8%
Software Multidrop PicoIT Software
Dimensions (L x W x H) 47 x 38 x 23 cm
Weight (English) 31.1 lb
Weight (Metric) 14.1 kg
Electrical Requirements 100 to 240V 50/60Hz
Voltage 100/240 V
Frequency 50/60 Hz
Mostrar más Mostrar menos
Do the Multidrop Pico 1 & Pico 8 Digital Dispensers come with a laptop?

No, Multidrop Pico 1 & Pico 8 Digital Dispensers are not sold with a laptop. This would need to be purchased separately. You can also use your own laptop or PC, as long as it meets the minimum PC requirements outlined in Chapter 3 of the Thermo Scientific Multidrop Pico 1 or Multidrop Pico 8 Operating Manuals:

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.