missing translation for 'onlineSavingsMsg'
Learn More

EKASTU Safety Multi-type Combined Filter 200 A1 B1 E1 K1-P3R D

Product Code. 10596433
Change view
Click to view available options
:
Multi-type Combined Filter 200 A1 B1 E1 K1-P3R D
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10596433 Multi-type Combined Filter 200 A1 B1 E1 K1-P3R D
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10596433 Supplier EKASTU Safety Supplier No. 422399

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

EKASTU Multi-type Combined Filter 200 A1 B1 E1 K1-P3R D offers protection against organic gases and vapours with a boiling point >65°C, inorganic gases and vapours, sulphur dioxide, ammonia and particles of toxic and highly toxic substances (P3)

  • 4 per pack
  • Innovative BIOSTOP
  • Dolomite clogging test

Specifications

Cartridge Type In connection with half masks Polimask 100/2, Polimask BETA, Polimask BETA/Silicone(DIN EN 140) and full mask C 607/TWIN (DIN EN 136)
Certifications/Compliance DIN EN 14387
Color Black
Disposable Reusable
Filter Type Multi-type Combined Filter
Material Sorbents: Impregnated active charcoal; Particle filter: Microfiberglass, Cellulose fibers, Addition (BIOSTOP)
Product Type 200 A1 B1 E1 K1-P3R D
Material (Housing) Polystyrene
Model Multi-type Combined Filter 200 A1 B1 E1 K1-P3R D
Dimensions (L x W x H) 16 x 16 x 9 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.