missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ Multi Tube Digital Vortex

Product Code. 17785019
Click to view available options
Plug Type:
US, EU, UK, AUS Plug
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17785019

Brand: Grant Instruments™ MSV3500

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The Grant Instruments MSV-3500 is a versatile multi speed vortex mixer designed for soft or intensive mixing of reagents in different size and type plastic tubes (0.2 to 50 mL).

  • Adjustable speed control: 300 to 3500 rpm
  • 4 mm orbit
  • Set and actual values of time and speed displayed
  • Acceleration time to max speed: 15 seconds
  • Speed and time under microprocessor control
  • LCD display indicates two lines of values: the set and actual values of speed and time

The MSV-3500 has four interchangeable platforms available (Platforms are sold separately or available as part of a kit (MSV-3500 4P):

  • SV-4/30 for 4 x 50 mL (30 mm) tubes
  • SV-10/10 for 10 x 10 mL (12 mm) tubes
  • SV-16/8 with 16/8/8 sockets for 1.5/0.5/0.2 mL microtubes
  • SV-8/15 for 8 x 15 mL (16 mm) tubes

Applications:

  • Designed for operation in lifescience laboratories working in the fields of biochemistry, cello and molecular biology.
  • Intensive stirring of bacterial and yeast cells, extraction of metabolites and enzymes from cells and cell cultures, and various DNA operations such as deproteinisation of DNA/protein complexes and purification of low molecular weight DNA/RNA fragments in PCR diagnostics.
TRUSTED_SUSTAINABILITY

Specifications

Orbit 4 mm
Load Bearing Capacity (Metric) 0.2 kg
Speed Range 300 to 3500 rpm
Voltage 12 V
Wattage 12 W
Amperage 1 A
Tube Size 1.5, 0.5, 0.2, 10, 15, 50 mL
Tube Capacity Up to 50 mL
Product Type Multi tube digital vortex
Plug Type US, EU, UK, AUS Plug
Dimensions (L x W x H) 32 x 23 x 24 cm
Shipping Weight (Metric) 3 kg
Weight (Metric) 2.6 kg
Color White
Model MSV-3500
Display LCD, 2 x 16 signs
Includes Platforms not included
Timer Range 0 to 60 min., Non-stop
Warranty and Service Offering 2 years
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.