missing translation for 'onlineSavingsMsg'
Learn More

Premiere Ecoforce™ Multi Purpose Cleaner Ready to Use Spray

Product Code. 15862815
Encompass_Preferred
Click to view available options
Volume (Metric):
750mL
This item is not returnable. View return policy

Product Code. 15862815

Brand: Premiere Ecoforce™ 11525

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Premiere Ecoforce™ Multi Purpose Cleaner Ready to Use Spray delivers outstanding cleaning results and have been independently awarded the Ecolabel, which is the official European environmental standard.

  • Ready to use high performance multi-purpose cleaner
  • Alkaline multi-purpose cleaner and degreaser
  • Suitable for cleaning most washable hard surfaces
  • Delivers outstanding eco-friendly cleaning
  • Use for damp wiping and spray cleaning applications
  • DO NOT use on terrazzo, aluminum or galvanised surfaces

Specifications

Product Type Multi-purpose Cleaner
pH 10
Boiling Point 100°C
For Use With (Application) Neutral detergent for removal of dirt from floors and hard surfaces
Quantity 6/Cs.
Volume (Metric) 750mL
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.