missing translation for 'onlineSavingsMsg'
Learn More

Premiere Ecoforce™ Multi Purpose Cleaner

Product Code. 15852805
Encompass_Preferred
Change view
Click to view available options
Volume (Metric):
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric)
15852805 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 15852805 Supplier Premiere Ecoforce™ Supplier No. 11500
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Premiere Ecoforce™ Multi Purpose Cleaner delivers outstanding cleaning results and have been independently awarded the Ecolabel, which is the official European environmental standard.

  • EU-Ecolabel accredited multi-purpose cleaner
  • High performance, concentrated neutral multi-purpose cleaner for the removal of dirt from floors and hard surfaces
  • Suitable for use on laminate, stainless steel and painted surfaces. Use for mopping, damp wiping, spray cleaning and deep cleaning applications

Specifications

Product Type Multi-purpose Cleaner
pH 7
Appearance Dark Orange
Boiling Point 100°C
Description Causes serious eye irritation
For Use With (Application) Neutral detergent for removal of dirt from floors and hard surfaces
Quantity 2 Pk.
Volume (Metric) 5 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.