missing translation for 'onlineSavingsMsg'
Learn More

Multi-Parameter Refractive Index & Density Certified Reference Material, at 15 °C, 20 °C and 25 °C, UKAS ISO 17025 / ISO 17034 Certified, Single 30 mL (Nominal RI Value 1.5463 at 20 °C), Paragon Scientific™

Product Code. 16080300
Click to view available options
Quantity:
30 mL
Unit Size:
30mL
This item is not returnable. View return policy

Product Code. 16080300

Brand: Paragon Scientific PSRVD03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Manufactured for refractive index and density measurements. The combined Refractive Index and Density CRM is available in four different materials, with each material including certified data for both refractive index and density at 15 °C, 20 °C and 25 °C.

  • Multi-Parameter Certified Reference Materials for Refractive Index and Density at 15 °C, 20 °C and 25 °C
  • Certified in strict accordance to ISO 17025 and ISO 17034 under our UKAS accreditation
  • Highest level of accreditation guarantee, providing the most credible certified data available worldwide
  • Density certified in accordance with primary level ASTM D1480 methodology
  • Ensures full compliance to ASTM and IP test method protocols
  • All measurements are fully traceable to NIST and international protocols
  • Low levels of uncertainty, ensuring maximum accuracy of data and dependable results
  • Supplied in 30 mL volume sealed glass vials for ample measurement readings
  • 12 month shelf life

Specifications

Quantity 30 mL

Nominal values quoted. Certified Values will vary from batch to batch.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.