missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Multi-Parameter Refractive Index & Density Certified Reference Material, at 15 °C, 20 °C and 25 °C, UKAS ISO 17025 / ISO 17034 Certified, Single 30 mL (Nominal RI Value 1.3330 at 20 °C), Paragon Scientific™
Click to view available options
Quantity:
30 mL
Unit Size:
30mL
Description
- Multi-Parameter Certified Reference Materials for Refractive Index and Density at 15 °C, 20 °C and 25 °C
- Certified in strict accordance to ISO 17025 and ISO 17034 under our UKAS accreditation
- Highest level of accreditation guarantee, providing the most credible certified data available worldwide
- Density certified in accordance with primary level ASTM D1480 methodology
- Ensures full compliance to ASTM and IP test method protocols
- All measurements are fully traceable to NIST and international protocols
- Low levels of uncertainty, ensuring maximum accuracy of data and dependable results
- Supplied in 30 mL volume sealed glass vials for ample measurement readings
- 12 month shelf life
Specifications
Specifications
| Quantity | 30 mL |
Nominal values quoted. Certified Values will vary from batch to batch.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur