missing translation for 'onlineSavingsMsg'
Learn More

Multi-Element QC-21 Check Standard Solution, Specpure™

Product Code. 11364038
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11364038

Brand: Thermo Scientific Alfa Aesar 041328.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Multi-Element QC-21 Check Standard Solution is used as a certified reference material or quality control check standard for inductively coupled plasma optical emission spectroscopy (ICP-OES). It is also used in inductively coupled plasma mass spectrometry (ICP-MS), and flame or furnace atomic absorption spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Multi-Element QC-21 Check Standard Solution
Physical Form Liquid
Concentration Matrix: 5% HNO3/tr. F-/tr. tartaric acid
MDL Number MFCD00803610
UN Number UN3264
Solubility Miscible with water
Quantity 100 mL
Grade Specpure

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.