Learn More
Multi-Element QC-21 Check Standard Solution, Specpure™
Description
Multi-Element QC-21 Check Standard Solution is used as a certified reference material or quality control check standard for inductively coupled plasma optical emission spectroscopy (ICP-OES). It is also used in inductively coupled plasma mass spectrometry (ICP-MS), and flame or furnace atomic absorption spectroscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Multi-Element QC-21 Check Standard Solution |
| Physical Form | Liquid |
| Concentration | Matrix: 5% HNO3/tr. F-/tr. tartaric acid |
| MDL Number | MFCD00803610 |
| UN Number | UN3264 |
| Solubility | Miscible with water |
| Quantity | 500 mL |
| Grade | Specpure |
Safety and Handling
- Multi-Element QC-21 Check Standard Solution, Specpure
- Danger
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contain: Nitric acid, Tartaric acid, Hydrofluoricacid
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.