Learn More
Multi-Element QC-19 Check Standard Solution, Specpure™, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 039146.AE
Code : NEW
Additional Details : Weight : 0.01000kg Transport : UN number : 3264 Chem class : 8 Pack group : III
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Multi-Element QC-19 Check Standard Solution | |
100 mL | |
UN3264 | |
Liquid |
Matrix: 5% HNO3/tr. F-/tr. tartaric acid | |
MFCD00210655 | |
Specpure™ |
RUO – Research Use Only