missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Multi-Element Organometallic Oil Standards 23 Element, 100μg/g; SPEX CertiPrep™
Shop All SPEX Certiprep Products
Click to view available options
Quantity:
50 g
100 g
Unit Size:
100g
50g
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Ag | 100μg/g |
| Al | 100μg/g |
| B | 100μg/g |
| Ba | 100μg/g |
| Ca | 100μg/g |
| Cd | 100μg/g |
| Cr | 100μg/g |
| Cu | 100μg/g |
| Fe | 100μg/g |
| K | 100μg/g |
| Mg | 100μg/g |
| Mn | 100μg/g |
| Mo | 100μg/g |
| Na | 100μg/g |
| Ni | 100μg/g |
| P | 100μg/g |
| Pb | 100μg/g |
| Sb | 100μg/g |
| Si | 100μg/g |
| Sn | 100μg/g |
| Ti | 100μg/g |
| V | 100μg/g |
| Zn | 100μg/g |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Chemical Name or Material | 23 Element Oil Standard |
| Packaging | Triple-leached LDPE bottle |
| Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
| Analyte or Component Names | Al,Ba,B |
| For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
| Type | Certified Reference Material |
| Element | Al, Ba, B |
| Solvent or Matrix | Base Oil 75 |
| Industry Type | Petrochemical Testing |
| Quantity | 100 g |
Product intended for laboratory use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering