Learn More
Multi-element Ionic Chromatography Cation Standard solution, H2O matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Brand: Fisher Chemical CM1774
Code : 04
Additional Details : Weight : 0.50000kg
Description
Multi-element Ionic Chromatography Cation Standard solution containing 4 components; 10mg/kg each of Lithium (Li+), Sodium (Na+), Potassium (K+), Calcium (Ca2+) for Ion Chromatography H2O matrixSpecifications
Four Components Solution | |
Bottle | |
Multi-component Cationic Standard Solution | |
H2O |
10mg/Kg of each component | |
500 mL | |
Lithium, Sodium, Potassium, Calcium | |
Liquid |